accessibility menu, dialog, popup

UserName

CAS RN 2557-77-9

IUPAC Name: 3-fluorobenzene-1-thiol
Synonyms: 3-fluorobenzenethiol
Molecular Weight (g/mol): 128.16
Molecular Formula: C6H5FS
InChi Key: ZDEUGINAVLMAET-UHFFFAOYSA-N
SMILES: FC1=CC=CC(S)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)