accessibility menu, dialog, popup

UserName

H3COSiOCH3

CAS RN 2553-19-7

IUPAC Name: diethoxydiphenylsilane
Synonyms: diphenyldiethoxysilane
Molecular Weight (g/mol): 272.42
Molecular Formula: C16H20O2Si
InChi Key: ZZNQQQWFKKTOSD-UHFFFAOYSA-N
SMILES: CCO[Si](OCC)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results