accessibility menu, dialog, popup

UserName

NClF

CAS RN 2546-56-7

IUPAC Name: 4-chloro-3-fluoropyridine
Synonyms: 4-chloro-3-fluoropyridine
Molecular Weight (g/mol): 131.53
Molecular Formula: C5H3ClFN
InChi Key: BEQUUSCRAKEKQM-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CN=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)