missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluoropyridine, 95%

Catalog No. AC443160250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443160250 25 g
AC443160050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443160250 Supplier Thermo Scientific Chemicals Supplier No. 443160250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2546-56-7 | C5H3ClFN | 131.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2546-56-7
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.53
MDL Number MFCD03453233
InChI Key BEQUUSCRAKEKQM-UHFFFAOYSA-N
Synonym pyridine, 4-chloro-3-fluoro, 3-fluoro-4-chloro pyridine, 3-fluoro-4-chloropyridine hydrochloride, pubchem6667, 4-chloro-3fluoropyridine, 3-fluoro-4-chloropyridine, acmc-209x6p, 4-chloro-3-fluoro-pyridin, 3-fluoro-4-chloro-pyridine, 4-chloro-3-fluoro-pyridine
PubChem CID 3852429
IUPAC Name 4-chloro-3-fluoropyridine
SMILES FC1=C(Cl)C=CN=C1

Specifications

Melting Point -27°C to 24°C
Boiling Point 142°C to 143°C
Flash Point 28°C
Assay Percent Range 94% min. (GC)
Quantity 25 g
Formula Weight 131.54
Percent Purity 95%
Chemical Name or Material 4-Chloro-3-fluoropyridine
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only