accessibility menu, dialog, popup

UserName

CH3H3COPOCH3H3COCH3CH3

CAS RN 25448-25-3

IUPAC Name: tris(8-methylnonyl) phosphite
Synonyms: phosphorous acid, triisodecyl ester
Molecular Weight (g/mol): 502.81
Molecular Formula: C30H63O3P
InChi Key: QEDNBHNWMHJNAB-UHFFFAOYSA-N
SMILES: CC(C)CCCCCCCOP(OCCCCCCCC(C)C)OCCCCCCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results