missing translation for 'onlineSavingsMsg'
Learn More

Triisodecyl Phosphite (mixture of isomers) 95.0+%, TCI America™

Catalog No. T042025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T042025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T042025ML Supplier TCI America Supplier No. T042025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25448-25-3
Molecular Formula C30H63O3P
Molecular Weight (g/mol) 502.805
MDL Number MFCD00015059
InChI Key QEDNBHNWMHJNAB-UHFFFAOYSA-N
Synonym Phosphorous Acid Triisodecyl Ester
PubChem CID 117488
IUPAC Name tris(8-methylnonyl) phosphite
SMILES CC(C)CCCCCCCOP(OCCCCCCCC(C)C)OCCCCCCCC(C)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 502.80
Physical Form Liquid
Chemical Name or Material Triisodecyl Phosphite (mixture of isomers)
EINECSNumber (2)-1894
RTECSNumber TH1140000
TSCA Yes