accessibility menu, dialog, popup

UserName

CAS RN 25387-93-3

IUPAC Name: lithium(1+) quinolin-8-olate
Synonyms: lithium(1+) quinolin-8-olate
Molecular Weight (g/mol): 151.09
Molecular Formula: C9H6LiNO
InChi Key: FQHFBFXXYOQXMN-UHFFFAOYSA-M
SMILES: [Li+].[O-]C1=C2N=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results