missing translation for 'onlineSavingsMsg'
Learn More

(8-Quinolinolato)lithium 98.0+%, TCI America™

Catalog No. Q01005G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q01005G 5 g
Q01001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q01005G Supplier TCI America Supplier No. Q01005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25387-93-3
Molecular Formula C9H6LiNO
Molecular Weight (g/mol) 151.093
MDL Number MFCD00152778
InChI Key FQHFBFXXYOQXMN-UHFFFAOYSA-M
PubChem CID 23686653
IUPAC Name lithium;quinolin-8-olate
SMILES [Li+].C1=CC2=C(C(=C1)[O-])N=CC=C2

Specifications

Melting Point 368°C
Color Yellow
Quantity 5 g
Formula Weight 151.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (8-Quinolinolato)lithium