accessibility menu, dialog, popup

UserName

NBrOBr

CAS RN 25373-69-7

IUPAC Name: 2,6-dibromopyridin-1-ium-1-olate
Synonyms: 2,6-dibromopyridin-1-ium-1-olate
Molecular Weight (g/mol): 252.89
Molecular Formula: C5H3Br2NO
InChi Key: YOJAFCXACCYASM-UHFFFAOYSA-N
SMILES: [O-][N+]1=C(Br)C=CC=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results