missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyridine N-Oxide 98.0+%, TCI America™

Catalog No. D50155G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D50155G Supplier TCI America Supplier No. D50155G
Only null left

Chemical Identifiers

CAS 25373-69-7
Molecular Formula C5H3Br2NO
Molecular Weight (g/mol) 252.893
MDL Number MFCD00234019
InChI Key YOJAFCXACCYASM-UHFFFAOYSA-N
PubChem CID 564873
IUPAC Name 2,6-dibromo-1-oxidopyridin-1-ium
SMILES C1=CC(=[N+](C(=C1)Br)[O-])Br

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 5 g
Formula Weight 252.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromopyridine N-Oxide