CAS RN 25309-65-3
IUPAC Name:
4-ethylbenzonitrile
Synonyms:
4-ethylbenzonitrile
Molecular Weight (g/mol):
131.18
Molecular Formula:
C9H9N
InChi Key:
SXFFMFAQNAFSLF-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
Promotions Available
4-Ethylbenzonitrile, 98%, Thermo Scientific Chemicals
4-Ethylbenzonitrile, 98%, Thermo Scientific Chemicals
CAS: 25309-65-3 Molecular Formula: C9H9N Molecular Weight (g/mol): 131.178 MDL Number: MFCD00016383 InChI Key: SXFFMFAQNAFSLF-UHFFFAOYSA-N Synonym: benzonitrile, 4-ethyl,p-ethylbenzonitrile,1-cyano-4-ethylbenzene,4-ethylbenzenecarbonitrile,4-ethyl-benzonitrile,acmc-20amj0,4-ethylbenzonitrile PubChem CID: 91393 IUPAC Name: 4-ethylbenzonitrile SMILES: CCC1=CC=C(C=C1)C#N