accessibility menu, dialog, popup

UserName

CAS RN 25300-37-2

IUPAC Name: 2-chloro-6-methylbenzene-1-sulfonyl chloride
Synonyms: 2-chloro-6-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 225.08
Molecular Formula: C7H6Cl2O2S
InChi Key: FJVZVTWCAGCLCS-UHFFFAOYSA-N
SMILES: CC1=C(C(Cl)=CC=C1)S(Cl)(=O)=O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)