CAS RN 251992-66-2
CAS RN 251992-66-2
IUPAC Name:
4-[(4-methylphenyl)sulfanyl]thieno[2,3-c]pyridine-2-carboxamide
Synonyms:
4-[(4-methylphenyl)sulfanyl]thieno[2,3-c]pyridine-2-carboxamide
Molecular Weight (g/mol):
300.39
Molecular Formula:
C15H12N2OS2
InChi Key:
QQGWEXFLMJGCAL-UHFFFAOYSA-N
SMILES:
CC1=CC=C(SC2=C3C=C(SC3=CN=C2)C(N)=O)C=C1
1
–
1
of
1
results
1
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide 95.0+%, TCI America™
CAS: 251992-66-2 Molecular Formula: C15H12N2OS2 Molecular Weight (g/mol): 300.394 MDL Number: MFCD09038566 InChI Key: QQGWEXFLMJGCAL-UHFFFAOYSA-N Synonym: 4-p-tolylthio thieno 2,3-c pyridine-2-carboxamide,unii-tp7tb1sszd,tp7tb1sszd,4-4-methylphenyl thio thieno 2,3-c pyridine-2-carboxamide,4-p-tolylsulfanyl thieno 2,3-c pyridine-2-carboxamide,thieno 2,3-c pyridine-2-carboxamide, 4-4-methylphenyl thio,4-4-methylphenyl thio thieno 2,3-c pyridine-2-ca,4-p-tolylsulfanyl-thieno 2,3-c pyridine-2-carboxylic acid amide PubChem CID: 9839311 IUPAC Name: 4-(4-methylphenyl)sulfanylthieno[2,3-c]pyridine-2-carboxamide SMILES: CC1=CC=C(C=C1)SC2=C3C=C(SC3=CN=C2)C(=O)N