missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 200°C |
| Color | White-Yellow |
| Quantity | 25 mg |
| Formula Weight | 300.39 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-(p-Tolylthio)thieno[2,3-c]pyridine-2-carboxamide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |