accessibility menu, dialog, popup

UserName

ClH3COONH3CCH3H3CCH3CH3H3CCH3

CAS RN 25155-18-4

IUPAC Name: benzyldimethyl(2-{2-[3-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
Synonyms: methylbenzethonium chloride 甲苄索氯铵
Molecular Weight (g/mol): 462.12
Molecular Formula: C28H44ClNO2
InChi Key: WYWSKWZUWZLELX-UHFFFAOYSA-M
SMILES: [Cl-].CC1=CC(OCCOCC[N+](C)(C)CC2=CC=CC=C2)=CC=C1C(C)(C)CC(C)(C)C
Physical Form 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)