accessibility menu, dialog, popup

UserName

CAS RN 25016-17-5

IUPAC Name: methyl 5-methyl-1H-pyrazole-3-carboxylate
Synonyms: methyl 5-methyl-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: GFEZEVUIYRGWNU-UHFFFAOYSA-N
SMILES: COC(=O)C1=NNC(C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results