missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-methyl[1H]pyrazole-3-carboxylate, 96%

Catalog No. AAH3613006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3613006 5 g
AAH3613003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3613006 Supplier Thermo Scientific Chemicals Supplier No. H3613006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25016-17-5 | C6H8N2O2 | 140.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25016-17-5
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD03778987
InChI Key GFEZEVUIYRGWNU-UHFFFAOYSA-N
Synonym methyl 3-methyl-1h-pyrazole-5-carboxylate, methyl 5-methylpyrazole-3-carboxylate, methyl 5-methyl-2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-methyl-, methyl ester, 5-methyl-1h-pyrazole-3-carboxylic acid methyl ester, 5-methyl-2h-pyrazole-3-carboxylic acid methyl ester, 5-methyl-1 2 h-pyrazole-3-carboxylic acid methyl ester, methylmethylpyrazolecarboxylate, ksc554q1l, 5-methoxycarbonyl-3-me-thylpyrazole
PubChem CID 2753079
IUPAC Name methyl 5-methyl-1H-pyrazole-3-carboxylate
SMILES CC1=CC(=NN1)C(=O)OC

Specifications

Melting Point 79°C to 81°C
Quantity 5 g
Formula Weight 140.14
Percent Purity 96%
Chemical Name or Material Methyl 5-methyl[1H]pyrazole-3-carboxylate
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only