accessibility menu, dialog, popup

UserName

BrOOOBrOOO

CAS RN 24848-78-0

IUPAC Name: dibromo-5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
Synonyms: dibromo-5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
Molecular Weight (g/mol): 375.91
Molecular Formula: C10Br2O6
InChi Key: OYIHYCLAXDDYFC-UHFFFAOYSA-N
SMILES: BrC1=C2C(=O)OC(=O)C2=C(Br)C2=C1C(=O)OC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results