missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibromopyromellitic Dianhydride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 275°C |
| Quantity | 5 g |
| Formula Weight | 375.91 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dibromopyromellitic Dianhydride |
Safety and Handling
| TSCA | No |