accessibility menu, dialog, popup

UserName

H3CBrCH3Br

CAS RN 24774-58-1

IUPAC Name: 2,5-dibromohexane
Synonyms: 2,5-dibromohexane
Molecular Weight (g/mol): 243.97
Molecular Formula: C6H12Br2
InChi Key: MQYLGFBWOZXHHF-UHFFFAOYNA-N
SMILES: CC(Br)CCC(C)Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results