accessibility menu, dialog, popup

UserName

CAS RN 2448-45-5

OHONHOO

CAS RN 2448-45-5

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-phenylpropanoic acid
Synonyms: carbobenzoxyphenylalanine
Molecular Weight (g/mol): 299.33
Molecular Formula: C17H17NO4
InChi Key: RRONHWAVOYADJL-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results