CAS RN 24468-13-1
IUPAC Name:
2-ethylhexyl carbonochloridate
Synonyms:
2-ethylhexyl chloroformate
Molecular Weight (g/mol):
192.68
Molecular Formula:
C9H17ClO2
InChi Key:
RTGLJCSUKOLTEM-UHFFFAOYNA-N
SMILES:
CCCCC(CC)COC(Cl)=O
1
–
1
of
1
results
1
2-Ethylhexyl Chloroformate 98.0+%, TCI America™
CAS: 24468-13-1 Molecular Formula: C9H17ClO2 Molecular Weight (g/mol): 192.683 MDL Number: MFCD00000643 InChI Key: RTGLJCSUKOLTEM-UHFFFAOYSA-N Synonym: Chloroformic Acid Octyl Ester, Chloroformic Acid 2-Ethylhexyl Ester, Octyl Chloroformate PubChem CID: 62783 IUPAC Name: 2-ethylhexyl carbonochloridate SMILES: CCCCC(CC)COC(=O)Cl