missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Chloroformate 98.0+%, TCI America™

Catalog No. C0927500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0927500ML 500 mL
C092725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0927500ML Supplier TCI America Supplier No. C0927500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24468-13-1
Molecular Formula C9H17ClO2
Molecular Weight (g/mol) 192.683
MDL Number MFCD00000643
InChI Key RTGLJCSUKOLTEM-UHFFFAOYSA-N
Synonym Chloroformic Acid Octyl Ester, Chloroformic Acid 2-Ethylhexyl Ester, Octyl Chloroformate
PubChem CID 62783
IUPAC Name 2-ethylhexyl carbonochloridate
SMILES CCCCC(CC)COC(=O)Cl

Specifications

Color Yellow
Boiling Point 107°C
Quantity 500 mL
UN Number 2748
Formula Weight 192.68
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Chloroformate
EINECSNumber (2)-1710
RTECSNumber FG3660000
TSCA Yes