accessibility menu, dialog, popup

UserName

CAS RN 2438-72-4

IUPAC Name: 2-(4-butoxyphenyl)-N-hydroxyacetamide
Synonyms: bufexamic acid
Molecular Weight (g/mol): 223.27
Molecular Formula: C12H17NO3
InChi Key: MXJWRABVEGLYDG-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(CC(=O)NO)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results