accessibility menu, dialog, popup

UserName

CAS RN 2430-27-5

IUPAC Name: 2-propylpentanamide
Synonyms: valpromide
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: OMOMUFTZPTXCHP-UHFFFAOYSA-N
SMILES: CCCC(CCC)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results