missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-propylacetamide, 97%

Catalog No. AAL0884714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0884714 25 g
AAL0884706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0884714 Supplier Thermo Scientific Chemicals Supplier No. L0884714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2430-27-5 | C8H17NO | 143.23 g/mol

It is employed as an antiepileptic drug. It is hydrolyzed quickly to VPA in vivo, but has intrinsic anticonvulsant activity. Also a reactant used in the synthesis of mutual prodrugs containing bio-cleavable and drug releasable disulfide linkers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform, dimethyl sulfoxide and methanol. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 2430-27-5
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD00051534
InChI Key OMOMUFTZPTXCHP-UHFFFAOYSA-N
Synonym valpromide, depamide, dipropylacetamide, valpramide, 2-propylpentamide, 2-propylvaleramide, pentanamide, 2-propyl, 2,2-di-n-propylacetamide, 2-ethylvaleramide, propyl-2 valeramide
PubChem CID 71113
ChEBI CHEBI:74562
IUPAC Name 2-propylpentanamide
SMILES CCCC(CCC)C(N)=O

Specifications

Melting Point 123°C to 127°C
Odor Odorless
Quantity 25 g
Beilstein 1750444
Merck Index 14,9914
Solubility Information Soluble in chloroform,dimethyl sulfoxide and methanol. Insoluble in water.
Formula Weight 143.23
Percent Purity 97%
Chemical Name or Material 2,2-Di-n-propylacetamide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 219-394-2
RTECSNumber YV5965500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only