accessibility menu, dialog, popup

UserName

NNH2OHO

CAS RN 24242-20-4

IUPAC Name: 5-aminopyridine-2-carboxylic acid
Synonyms: 5-aminopicolinic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: WDJARUKOMOGTHA-UHFFFAOYSA-N
SMILES: NC1=CN=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)