missing translation for 'onlineSavingsMsg'
Learn More

5-Aminopyridine-2-carboxylic acid, 95%

Catalog No. AAH2758906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2758906 5 g
AAH2758903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2758906 Supplier Thermo Scientific Chemicals Supplier No. H2758906

CAS: 24242-20-4 | C6H6N2O2 | 138.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24242-20-4
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD02684600
InChI Key WDJARUKOMOGTHA-UHFFFAOYSA-N
Synonym 5-amino-2-pyridinecarboxylic acid, 5-aminopicolinic acid, 5-amino-2-picolinic acid, 2-pyridinecarboxylic acid, 5-amino, 5-amino-pyridine-2-carboxylic acid, 5-amino-2-pyridinecarboxylicacid, pubchem15896, 5-amino-2-carboxypyridine, acmc-209ga3, ksc201o9n
PubChem CID 4179726
IUPAC Name 5-aminopyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1N)C(=O)O

Specifications

Melting Point 217°C to 219°C
Odor Odorless
Quantity 5 g
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 5-Aminopyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only