CAS RN 2413-38-9
IUPAC Name:
dihydrogen 2-(4-{3-[(9Z)-2-(trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl}piperazin-1-yl)ethan-1-ol dichloride
Synonyms:
dihydrogen α-flupenthixol dichloride
Molecular Weight (g/mol):
507.44
Molecular Formula:
C23H27Cl2F3N2OS
InChi Key:
IOVDQEIIMOZNNA-MHKBYHAFSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].OCCN1CCN(CC\C=C2\C3=CC=CC=C3SC3=CC=C(C=C23)C(F)(F)F)CC1
1
–
2
of
2
results
1
Promotions Available
Flupentixol dihydrochloride, Thermo Scientific Chemicals
Flupentixol dihydrochloride, Thermo Scientific Chemicals
CAS: 2413-38-9 Molecular Weight (g/mol): 507.44 InChI Key: IOVDQEIIMOZNNA-MHKBYHAFSA-N IUPAC Name: dihydrogen 2-(4-{3-[(9Z)-2-(trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl}piperazin-1-yl)ethan-1-ol dichloride SMILES: [H+].[H+].[Cl-].[Cl-].OCCN1CCN(CC\C=C2\C3=CC=CC=C3SC3=CC=C(C=C23)C(F)(F)F)CC1
2
R&D Systems™ Flupenthixol dihydrochloride
CAS: 2413-38-9 Molecular Formula: C23H27Cl2F3N2OS