missing translation for 'onlineSavingsMsg'
Learn More

Flupentixol dihydrochloride, Thermo Scientific Chemicals

Catalog No. AC464822500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464822500 250 mg
AC464820010 1 g
AC464820050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC464822500 Supplier Thermo Scientific Chemicals Supplier No. 464822500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2413-38-9
Molecular Weight (g/mol) 507.44
InChI Key IOVDQEIIMOZNNA-MHKBYHAFSA-N
IUPAC Name dihydrogen 2-(4-{3-[(9Z)-2-(trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl}piperazin-1-yl)ethan-1-ol dichloride
SMILES [H+].[H+].[Cl-].[Cl-].OCCN1CCN(CC\C=C2\C3=CC=CC=C3SC3=CC=C(C=C23)C(F)(F)F)CC1

Specifications

Molecular Formula C23H27Cl2F3N2OS
Quantity 250 mg
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: insoluble in ethyl acetate, methanol
Formula Weight 507.44
Chemical Name or Material Flupentixol dihydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 219-321-4

RUO – Research Use Only