accessibility menu, dialog, popup

UserName

CAS RN 2390-74-1

IUPAC Name: 2-(2-aminoacetamido)-3-(1H-indol-3-yl)propanoic acid
Synonyms: 2-(2-aminoacetamido)-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol): 261.28
Molecular Formula: C13H15N3O3
InChi Key: AJHCSUXXECOXOY-UHFFFAOYNA-N
SMILES: NCC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results