accessibility menu, dialog, popup

UserName

H3CMgCl

CAS RN 2386-64-3

IUPAC Name: chloro(ethyl)magnesium
Synonyms: ethylmagnesium chloride|chloroethylmagnesium
Molecular Weight (g/mol): 88.82
Molecular Formula: C2H5ClMg
InChi Key: ZWNJJACKPNTXHY-UHFFFAOYSA-M
SMILES: CC[Mg]Cl
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)