missing translation for 'onlineSavingsMsg'
Learn More

Ethylmagnesium Chloride (ca. 1.0mol/L in Tetrahydrofuran) activated with Zinc Chloride (ca. 10mol%), TCI America™

Catalog No. E0778250G
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0778250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E0778250G Supplier TCI America Supplier No. E0778250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2386-64-3
Molecular Formula C2H5ClMg
Molecular Weight (g/mol) 88.82
MDL Number MFCD00000472
InChI Key ZWNJJACKPNTXHY-UHFFFAOYSA-M
Synonym ethylmagnesium chloride, chloroethylmagnesium, ethylmagnesiumchloride, magnesium, chloroethyl, etmgcl, chloro ethyl magnesium, ethyl magnesium chloride, chloroethyl-magnesiu, ethyl magnesiumchloride, ethylmagnesium chloride solution
PubChem CID 75432
IUPAC Name chloro(ethyl)magnesium
SMILES CC[Mg]Cl

Specifications

Quantity 250 g
UN Number 3399
Formula Weight 88.82
Chemical Name or Material Ethylmagnesium Chloride (ca. 1.0mol/L in Tetrahydrofuran) activated with Zinc Chloride (ca. 10mol%)