accessibility menu, dialog, popup

UserName

CH3SOOO(S)OH3CCH3O

CAS RN 23735-43-5

IUPAC Name: [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzene-1-sulfonate
Synonyms: [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 286.34
Molecular Formula: C13H18O5S
InChi Key: SRKDUHUULIWXFT-NSHDSACASA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H]1COC(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results