missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. D25505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25505G 5 g
D25501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25505G Supplier TCI America Supplier No. D25505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23735-43-5
Molecular Formula C13H18O5S
Molecular Weight (g/mol) 286.342
MDL Number MFCD00063234
InChI Key SRKDUHUULIWXFT-NSHDSACASA-N
Synonym s-+-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, s-+-2,2-dimethyl-1,3-dioxolane-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate, r---2,2-dimethyl-1,3-dioxolane-4-ylmethyl-p-toluenesulfonate, pubchem6795, 2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate #, s-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate
PubChem CID 2723760
IUPAC Name [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2COC(O2)(C)C

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 286.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator