accessibility menu, dialog, popup

UserName

NaNaNaOHNHNSOOOOSOOOSOOO(E)

CAS RN 23647-14-5

IUPAC Name: trisodium (3E)-5-hydroxy-4-oxo-3-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: trisodium (3E)-5-hydroxy-4-oxo-3-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 570.4
Molecular Formula: C16H9N2Na3O11S3
InChi Key: LSAWBVPBZOEGOT-ZOVWSQMSSA-K
SMILES: [Na+].[Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)S([O-])(=O)=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (5)
Grade 
  • (3)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)
  • (2)