missing translation for 'onlineSavingsMsg'
Learn More

Trisodium 2-(4-Sulfophenylazo)-1,8-dihydroxynaphthalene-3,6-disulfonate Hydrate 95.0+%, TCI America™

Catalog No. D06045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06045G 5 g
D060425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06045G Supplier TCI America Supplier No. D06045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23647-14-5
Molecular Formula C16H10N2Na3O11S3+
Molecular Weight (g/mol) 571.408
MDL Number MFCD00003952
InChI Key LSAWBVPBZOEGOT-UHFFFAOYSA-L
Synonym spadns, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-4-sulfophenyl azo-, trisodium salt, trisodium 4,5-dihydroxy-3-e-2-4-sulfonatophenyl diazen-1-yl naphthalene-2,7-disulfonate, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-2-4-sulfophenyl diazenyl-, sodium salt 1:3, 2-p-sulfophenylazo-1,8-dihydroxy-3,6-naph-thalenedisulfonic acid
PubChem CID 73658865
IUPAC Name trisodium;5-hydroxy-4-oxo-3-[(4-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O.[Na+].[Na+].[Na+]

Specifications

Quantity 5 g
Formula Weight 570.40
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trisodium 2-(4-Sulfophenylazo)-1,8-dihydroxynaphthalene-3,6-disulfonate Hydrate