accessibility menu, dialog, popup

UserName

ClOClO

CAS RN 2351-37-3

IUPAC Name: [1,1'-biphenyl]-4,4'-dicarbonyl dichloride
Synonyms: [1,1'-biphenyl]-4,4'-dicarbonyl dichloride
Molecular Weight (g/mol): 279.12
Molecular Formula: C14H8Cl2O2
InChi Key: QDBOAKPEXMMQFO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(Cl)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results