missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Biphenyldicarbonyl Chloride 97.0+%, TCI America™

Catalog No. B130810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B130810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B130810G Supplier TCI America Supplier No. B130810G
Only null left

Chemical Identifiers

CAS 2351-37-3
Molecular Formula C14H8Cl2O2
Molecular Weight (g/mol) 279.12
MDL Number MFCD00058934
InChI Key QDBOAKPEXMMQFO-UHFFFAOYSA-N
Synonym 4,4′C-Bibenzoyl Chloride, 4,4′C-Dibenzoyl Chloride, 4,4′C-Diphenyldicarbonyl Chloride
PubChem CID 75374
IUPAC Name [1,1'-biphenyl]-4,4'-dicarbonyl dichloride
SMILES ClC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 10 g
UN Number 3261
Formula Weight 279.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldicarbonyl Chloride