accessibility menu, dialog, popup

UserName

CAS RN 235097-76-4

H3CCH3OOSOOH3C

CAS RN 235097-76-4

IUPAC Name: 2-(propan-2-yloxy)ethyl methanesulfonate
Synonyms: 2-isopropoxyethyl methanesulfonate
Molecular Weight (g/mol): 182.23
Molecular Formula: C6H14O4S
InChi Key: YDKQNGZSQDGFRB-UHFFFAOYSA-N
SMILES: CC(C)OCCOS(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results