missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyethyl Methanesulfonate 98.0+%, TCI America™

Catalog No. M129725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M129725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M129725G Supplier TCI America Supplier No. M129725G
Only null left

Chemical Identifiers

CAS 235097-76-4
Molecular Formula C6H14O4S
Molecular Weight (g/mol) 182.234
MDL Number MFCD02093478
InChI Key YDKQNGZSQDGFRB-UHFFFAOYSA-N
Synonym Methanesulfonic Acid 2-Isopropoxyethyl Ester
PubChem CID 18465045
IUPAC Name 2-propan-2-yloxyethyl methanesulfonate
SMILES CC(C)OCCOS(=O)(=O)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 182.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropoxyethyl Methanesulfonate