accessibility menu, dialog, popup

UserName

CAS RN 233770-01-9

BrNI

CAS RN 233770-01-9

IUPAC Name: 3-bromo-5-iodopyridine
Synonyms: 3-bromo-5-iodopyridine
Molecular Weight (g/mol): 283.89
Molecular Formula: C5H3BrIN
InChi Key: AOOZLVWDZUPEHT-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(I)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)