missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-iodopyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 133°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | 3-bromo-5-iodo-pyridine, 5-bromo-3-iodopyridine, pyridine, 3-bromo-5-iodo, 3-bromo-5-iodpyridine, 3-iodo-5-bromopyridine, pubchem8080, acmc-1cdm9, pyridine,3-bromo-5-iodo, 3-bromo-5-iodopyridine, abbypharma ap-13-5787 |
| Formula Weight | 283.89 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Bromo-5-iodopyridine |
Safety and Handling
| TSCA | No |