CAS RN 23326-27-4
IUPAC Name:
methyl but-2-ynoate
Synonyms:
methyl but-2-ynoate
Molecular Weight (g/mol):
98.1
Molecular Formula:
C5H6O2
InChi Key:
UJQCANQILFWSDJ-UHFFFAOYSA-N
SMILES:
COC(=O)C#CC
1
–
2
of
2
results
1
Promotions Available
Methyl 2-butynoate, 97%
Methyl 2-butynoate, 97%
CAS: 23326-27-4 Molecular Formula: C5H6O2 Molecular Weight (g/mol): 98.1 MDL Number: MFCD00009273 InChI Key: UJQCANQILFWSDJ-UHFFFAOYSA-N Synonym: methyl 2-butynoate,methyl tetrolate,2-butynoic acid, methyl ester,2-butynoic acid methyl ester,tetrolic acid methyl ester,but-2-ynoic acid methyl ester,methyl2-butynoate,methyl 2-butynoate ?,pubchem12783,acmc-1ceil PubChem CID: 141019 IUPAC Name: methyl but-2-ynoate SMILES: CC#CC(=O)OC
2
Methyl Tetrolate 97.0+%, TCI America™
CAS: 23326-27-4 Molecular Formula: C5H6O2 Molecular Weight (g/mol): 98.101 MDL Number: MFCD00009273 InChI Key: UJQCANQILFWSDJ-UHFFFAOYSA-N PubChem CID: 141019 IUPAC Name: methyl but-2-ynoate SMILES: CC#CC(=O)OC