missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-butynoate, 97%

Catalog No. AC223490250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223490250 25 g Glass bottle
AC223490010 1 g Glass bottle
AC223490050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC223490250 Supplier Thermo Scientific Chemicals Supplier No. 223490250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23326-27-4 | C5H6O2 | 98.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23326-27-4
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
MDL Number MFCD00009273
InChI Key UJQCANQILFWSDJ-UHFFFAOYSA-N
Synonym methyl 2-butynoate, methyl tetrolate, 2-butynoic acid, methyl ester, 2-butynoic acid methyl ester, tetrolic acid methyl ester, but-2-ynoic acid methyl ester, methyl2-butynoate, methyl 2-butynoate ?, pubchem12783, acmc-1ceil
PubChem CID 141019
IUPAC Name methyl but-2-ynoate
SMILES CC#CC(=O)OC

Specifications

Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 76°C to 77°C (80.0 mmHg)
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4360 to 1.438
Linear Formula CH3C∽CCO2CH3
Quantity 25 g
Beilstein 02, III, 1449
Specific Gravity 0.98
Formula Weight 98.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 2-butynoate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only