accessibility menu, dialog, popup

UserName

CAS RN 2315-81-3

IUPAC Name: 3-chloro-4-hydroxybenzonitrile
Synonyms: 3-chloro-4-hydroxybenzonitrile
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: CRYPJUOSZDQWJZ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1Cl)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)