missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxybenzonitrile 98.0+%, TCI America™

Catalog No. C30835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30835G 5 g
C30831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30835G Supplier TCI America Supplier No. C30835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2315-81-3
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.565
MDL Number MFCD01567246
InChI Key CRYPJUOSZDQWJZ-UHFFFAOYSA-N
Synonym benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94
PubChem CID 2735739
IUPAC Name 3-chloro-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1C#N)Cl)O

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 5 g
Formula Weight 153.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-4-hydroxybenzonitrile