accessibility menu, dialog, popup

UserName

CAS RN 23138-55-8

IUPAC Name: 1-bromo-3-isocyanatobenzene
Synonyms: 1-bromo-3-isocyanatobenzene
Molecular Weight (g/mol): 198.02
Molecular Formula: C7H4BrNO
InChi Key: VQVBCZQTXSHJGF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)N=C=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)