missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenyl isocyanate, 97%

Catalog No. AC314130050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC314130050 5 g
AC314130010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314130050 Supplier Thermo Scientific Chemicals Supplier No. 314130050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23138-55-8 | C7H4BrNO | 198.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23138-55-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00002014
InChI Key VQVBCZQTXSHJGF-UHFFFAOYSA-N
Synonym 3-bromophenyl isocyanate, 3-bromophenylisocyanate, benzene, 1-bromo-3-isocyanato, isocyanic acid 3-bromophenyl ester, 3-bromobenzenisocyanate, m-bromophenylisocyanate, 3-bromophenyliso-cyanate, 3-bromo phenyl isocyanate, 3-bromo-phenyl isocyanate, acmc-209g1f
PubChem CID 141001
IUPAC Name 1-bromo-3-isocyanatobenzene
SMILES BrC1=CC=CC(=C1)N=C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5860g/mL
Boiling Point 220°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5835 to 1.5865
Linear Formula BrC6H4NCO
Quantity 5 g
Specific Gravity 1.586
Formula Weight 198.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromophenyl isocyanate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only