accessibility menu, dialog, popup

UserName

BrNHN

CAS RN 2302-25-2

IUPAC Name: 5-bromo-1H-imidazole
Synonyms: 4-bromo-1H-imidazole
Molecular Weight (g/mol): 146.98
Molecular Formula: C3H3BrN2
InChi Key: FHZALEJIENDROK-UHFFFAOYSA-N
SMILES: BrC1=CN=CN1
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)